Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: surf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201127.1 | NP_957421.1 | 2 - 3 | 360 | 80 | |
GenBank mRNA | BC056295.1 | - | 2 - 3 | 360 | 80 | |
BC066509.1 | - | 2 - 3 | 346 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |