Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dldh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201506.1 | NP_958914.1 | 6 - 7 | 508 | 83 | |
GenBank mRNA | AY391421.1 | - | 6 - 7 | 459 | 83 | |
BC044432.1 | - | 6 - 7 | 508 | 83 | ||
BC065616.1 | - | 6 - 7 | 485 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |