Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ewsr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212630.1 | NP_997795.1 | 9 - 10 | 941 | 98 | |
XM_005165107.2 | XP_005165164.2 | 9 - 10 | 928 | 98 | ||
XM_005165108.2 | XP_005165165.2 | 9 - 10 | 900 | 98 | ||
GenBank mRNA | BC044518.1 | - | 9 - 10 | 941 | 98 | |
BC067661.1 | - | 8 - 9 | 895 | 98 | ||
BC097019.1 | - | 9 - 10 | 934 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |