Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212677.1 | NP_997842.1 | 3 - 4 | 248 | 86 | |
GenBank mRNA | BC047848.1 | - | 3 - 4 | 248 | 86 | |
BC067159.1 | - | 3 - 4 | 240 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |