Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: impdh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002177.1 | NP_001002177.1 | 9 - 10 | 1113 | 85 | |
XM_005159007.4 | XP_005159064.1 | 9 - 10 | 1659 | 85 | ||
XM_005159008.4 | XP_005159065.1 | 9 - 10 | 1661 | 85 | ||
XM_009293441.2 | XP_009291716.1 | 9 - 10 | 1148 | 85 | ||
GenBank mRNA | BC074090.1 | - | 9 - 10 | 1113 | 85 | |
BC165226.1 | - | 8 - 9 | 943 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |