Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002333.1 | NP_001002333.1 | 3 - 4 | 392 | 141 | |
GenBank mRNA | BC076467.1 | - | 3 - 4 | 392 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |