Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsmce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002551.1 | NP_001002551.1 | 3 - 4 | 313 | 64 | |
GenBank mRNA | BC076252.1 | - | 3 - 4 | 313 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |