Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsmce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002551.1 | NP_001002551.1 | 6 - 7 | 699 | 76 | |
XM_005164207.3 | XP_005164264.1 | 7 - 8 | 825 | 76 | ||
GenBank mRNA | BC076252.1 | - | 6 - 7 | 699 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |