Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002671.1 | NP_001002671.1 | 2 - 3 | 613 | 66 | |
GenBank mRNA | BC076443.1 | - | 2 - 3 | 613 | 66 | |
BC165064.1 | - | 2 - 3 | 467 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |