Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csdc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002690.1 | NP_001002690.1 | 3 - 4 | 511 | 92 | |
GenBank mRNA | BC076447.1 | - | 3 - 4 | 511 | 92 | |
BC164903.1 | - | 2 - 3 | 327 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |