Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003416.2 | NP_001003416.2 | 2 - 3 | 422 | 82 | |
GenBank mRNA | AY648741.1 | - | 2 - 3 | 82 | ||
BC092743.1 | - | 2 - 3 | 422 | 82 | ||
BC165088.1 | - | 2 - 3 | 362 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |