Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhfpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003444.1 | NP_001003444.1 | 2 - 3 | 643 | 79 | |
GenBank mRNA | BC076060.1 | - | 2 - 3 | 643 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |