Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: htra3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003502.1 | NP_001003502.1 | 2 - 3 | 668 | 68 | |
GenBank mRNA | BC078402.1 | - | 2 - 3 | 668 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |