Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003991.1 | NP_001003991.1 | 9 - 10 | 914 | 73 | |
GenBank mRNA | BC080262.1 | - | 9 - 10 | 914 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |