Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gypc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005394.1 | NP_001005394.1 | 3 - 4 | 261 | 84 | |
GenBank mRNA | AY234186.1 | - | 3 - 4 | 261 | 84 | |
BC171465.1 | - | 2 - 3 | 93 | 84 | ||
BC171467.1 | - | 2 - 3 | 93 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |