Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005955.1 | NP_001005955.1 | 4 - 5 | 416 | 62 | |
XM_005166077.4 | XP_005166134.1 | 4 - 5 | 495 | 62 | ||
XM_005166078.4 | XP_005166135.1 | 4 - 5 | 494 | 62 | ||
GenBank mRNA | BC083444.1 | - | 4 - 5 | 416 | 62 | |
BC152157.1 | - | 4 - 5 | 416 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |