Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005955.1 | NP_001005955.1 | 6 - 7 | 524 | 90 | |
GenBank mRNA | BC083444.1 | - | 6 - 7 | 524 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |