Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ext1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012368.2 | NP_001012368.2 | 5 - 6 | 1937 | 69 | |
GenBank mRNA | AY734455.1 | - | 5 - 6 | 1368 | 69 | |
BC133117.1 | - | 5 - 6 | 1937 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |