Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012379.1 | NP_001012379.1 | 1 - 2 | 30 | 70 | |
GenBank mRNA | AB195468.1 | - | 1 - 2 | 30 | 70 | |
BC163126.1 | - | 1 - 2 | 115 | 70 | ||
BC163136.1 | - | 1 - 2 | 112 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |