Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03115945_g1
See other ABLIM1B GE Assays ›
Gene Symbol:
ablim1b
Gene Name:
actin binding LIM protein 1b
Gene Aliases:
si:dkeyp-84e12.1, zgc:112954
Chromosome Location:
Chr.12: 4690 - 44097 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03115945_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ablim1b
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_009306613.3
XP_009304888.1
5 - 6
956
72
XM_009306617.3
XP_009304892.1
5 - 6
954
72
XM_009306619.3
XP_009304894.1
5 - 6
953
72
XM_009306622.3
XP_009304897.1
5 - 6
952
72
XM_009306623.3
XP_009304898.1
5 - 6
951
72
XM_009306624.3
XP_009304899.1
5 - 6
953
72
XM_009306627.3
XP_009304902.1
5 - 6
953
72
XM_017358455.2
XP_017213944.1
5 - 6
954
72
XM_017358456.2
XP_017213945.1
5 - 6
952
72
XM_021480065.1
XP_021335740.1
5 - 6
955
72
XM_021480066.1
XP_021335741.1
5 - 6
956
72
XM_021480067.1
XP_021335742.1
5 - 6
955
72
XM_021480068.1
XP_021335743.1
5 - 6
955
72
XM_021480069.1
XP_021335744.1
5 - 6
957
72
XM_021480070.1
XP_021335745.1
5 - 6
956
72
XM_021480071.1
XP_021335746.1
5 - 6
955
72
XM_021480072.1
XP_021335747.1
5 - 6
956
72
XM_021480073.1
XP_021335748.1
5 - 6
955
72
XM_021480074.1
XP_021335749.1
5 - 6
955
72
XM_021480075.1
XP_021335750.1
5 - 6
955
72
XM_021480076.1
XP_021335751.1
5 - 6
955
72
XM_021480078.1
XP_021335753.1
5 - 6
956
72
XM_021480079.1
XP_021335754.1
5 - 6
956
72
XM_021480080.1
XP_021335755.1
5 - 6
957
72
XM_021480081.1
XP_021335756.1
5 - 6
955
72
XM_021480082.1
XP_021335757.1
5 - 6
954
72
XM_021480083.1
XP_021335758.1
5 - 6
955
72
XM_021480084.1
XP_021335759.1
5 - 6
954
72
XM_021480085.1
XP_021335760.1
5 - 6
953
72
XM_021480086.1
XP_021335761.1
5 - 6
876
72
XM_021480087.1
XP_021335762.1
5 - 6
747
72
XM_021480088.1
XP_021335763.1
5 - 6
956
72
XM_021480089.1
XP_021335764.1
5 - 6
958
72
XM_021480090.1
XP_021335765.1
5 - 6
956
72
XM_021480091.1
XP_021335766.1
5 - 6
958
72
XM_021480092.1
XP_021335767.1
5 - 6
956
72
XM_021480093.1
XP_021335768.1
5 - 6
955
72
XM_021480094.1
XP_021335769.1
5 - 6
954
72
XM_021480095.1
XP_021335770.1
5 - 6
955
72
XM_021480096.1
XP_021335771.1
5 - 6
954
72
XM_021480097.1
XP_021335772.1
5 - 6
954
72
XM_021480098.1
XP_021335773.1
5 - 6
953
72
XM_021480099.1
XP_021335774.1
5 - 6
963
72
XM_021480100.1
XP_021335775.1
5 - 6
963
72
XM_021480101.1
XP_021335776.1
5 - 6
962
72
XM_021480102.1
XP_021335777.1
5 - 6
962
72
XM_021480104.1
XP_021335779.1
5 - 6
962
72
GenBank mRNA
BC091932.1
-
5 - 6
785
72
EB891787.1
-
1 - 2
60
72
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Mammalian Gene Collection
Detect Genomic DNA
Danio rerio
Probe spans exons
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
lamellipodium assembly
cilium morphogenesis
actin binding
metal ion binding
actin filament binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0