Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pi4kaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034972.1 | NP_001030144.1 | 9 - 10 | 1006 | 70 | |
XM_005172596.4 | XP_005172653.1 | 10 - 11 | 1234 | 70 | ||
XM_009305081.3 | XP_009303356.1 | 10 - 11 | 1235 | 70 | ||
GenBank mRNA | AY929292.1 | - | 9 - 10 | 1006 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |