Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044937.2 | NP_001038402.1 | 11 - 12 | 1876 | 78 | |
XM_005170050.4 | XP_005170107.1 | 12 - 13 | 2556 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |