Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044937.2 | NP_001038402.1 | 2 - 3 | 935 | 69 | |
XM_005170050.4 | XP_005170107.1 | 3 - 4 | 1615 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |