Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044937.2 | NP_001038402.1 | 6 - 7 | 1418 | 86 | |
XM_005170050.4 | XP_005170107.1 | 7 - 8 | 2098 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |