Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slco3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045188.2 | NP_001038653.1 | 5 - 6 | 1125 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |