Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076637.1 | NP_001070105.1 | 8 - 9 | 1248 | 71 | |
XM_021481062.1 | XP_021336737.1 | 8 - 9 | 1207 | 71 | ||
XM_021481063.1 | XP_021336738.1 | 8 - 9 | 1209 | 71 | ||
XM_021481064.1 | XP_021336739.1 | 8 - 9 | 1301 | 71 | ||
XM_021481065.1 | XP_021336740.1 | 8 - 9 | 1298 | 71 | ||
GenBank mRNA | BC124221.1 | - | 8 - 9 | 1248 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |