Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdzrn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082923.2 | NP_001076392.1 | 6 - 7 | 678 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |