Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loxl5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102574.1 | NP_001096044.1 | 1 - 2 | 716 | 64 | |
XM_017357369.2 | XP_017212858.1 | 1 - 2 | 868 | 64 | ||
XM_017357372.2 | XP_017212861.1 | 2 - 3 | 1333 | 64 | ||
GenBank mRNA | EF030486.1 | - | 1 - 2 | 716 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |