Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313682.1 | NP_001300611.1 | 4 - 5 | 776 | 60 | |
XM_021475995.1 | XP_021331670.1 | 4 - 5 | 475 | 60 | ||
GenBank mRNA | AF152607.1 | - | 4 - 5 | 529 | 60 | |
AF179407.1 | - | 4 - 5 | 772 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |