Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr2f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131186.1 | NP_571261.1 | 1 - 2 | 1019 | 70 | |
GenBank mRNA | X70300.1 | - | 1019 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |