Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p2rx3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131623.1 | NP_571698.1 | 7 - 8 | 832 | 105 | |
GenBank mRNA | AF237683.1 | - | 7 - 8 | 838 | 105 | |
AF238292.1 | - | 7 - 8 | 832 | 105 | ||
BC162279.1 | - | 7 - 8 | 818 | 105 | ||
BC162600.1 | - | 7 - 8 | 818 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |