Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152973.2 | NP_694505.2 | 2 - 3 | 729 | 90 | |
XM_021469720.1 | XP_021325395.1 | 2 - 3 | 842 | 90 | ||
GenBank mRNA | AF397015.1 | - | 2 - 3 | 720 | 90 | |
BC061708.1 | - | 2 - 3 | 729 | 90 | ||
BC065358.1 | - | 2 - 3 | 723 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |