Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197933.2 | NP_922915.2 | 3 - 4 | 250 | 79 | |
GenBank mRNA | AF387900.1 | - | 3 - 4 | 297 | 79 | |
BC044507.1 | - | 3 - 4 | 250 | 79 | ||
BC165811.1 | - | 3 - 4 | 258 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |