Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198143.1 | NP_937786.1 | 1 - 2 | 102 | 106 | |
XM_005166666.4 | XP_005166723.1 | 2 - 3 | 247 | 106 | ||
GenBank mRNA | AF385081.1 | - | 1 - 2 | 102 | 106 | |
AY561516.1 | - | 2 - 3 | 146 | 106 | ||
BC075977.1 | - | 2 - 3 | 133 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |