Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199555.2 | NP_955849.2 | 4 - 5 | 670 | 62 | |
GenBank mRNA | AY398358.1 | - | 4 - 5 | 670 | 62 | |
BC045917.1 | - | 4 - 5 | 534 | 62 | ||
BC067193.1 | - | 4 - 5 | 497 | 62 | ||
BC165307.1 | - | 3 - 4 | 501 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |