Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itm2ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199646.1 | NP_955940.1 | 1 - 2 | 178 | 76 | |
GenBank mRNA | BC044480.1 | - | 1 - 2 | 178 | 76 | |
BC128819.1 | - | 1 - 2 | 154 | 76 | ||
BC164594.1 | - | 1 - 2 | 123 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |