Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adam17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199673.1 | NP_955967.1 | 11 - 12 | 1617 | 63 | |
XM_005158718.4 | XP_005158775.1 | 11 - 12 | 1777 | 63 | ||
GenBank mRNA | BC057410.1 | - | 11 - 12 | 1617 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |