Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2r2ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309438.1 | NP_001296367.1 | 8 - 9 | 1209 | 78 | |
GenBank mRNA | BC056824.1 | - | 7 - 8 | 892 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |