Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc1a2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199979.2 | NP_956273.2 | 8 - 9 | 1501 | 87 | |
XM_009298220.3 | XP_009296495.1 | 8 - 9 | 1286 | 87 | ||
XM_009298221.3 | XP_009296496.1 | 8 - 9 | 1265 | 87 | ||
XM_009298222.3 | XP_009296497.1 | 8 - 9 | 1452 | 87 | ||
GenBank mRNA | BC056751.1 | - | 8 - 9 | 1426 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |