Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328573.1 | NP_001315502.1 | 7 - 8 | 939 | 97 | |
NM_200015.2 | NP_956309.2 | 7 - 8 | 942 | 97 | ||
GenBank mRNA | BC047167.1 | - | 7 - 8 | 882 | 97 | |
BC065630.1 | - | 7 - 8 | 871 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |