Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200281.1 | NP_956575.1 | 3 - 4 | 356 | 137 | |
XM_021469763.1 | XP_021325438.1 | 3 - 4 | 306 | 137 | ||
GenBank mRNA | AY398411.1 | - | 3 - 4 | 393 | 137 | |
BC049345.1 | - | 3 - 4 | 356 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |