Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200353.2 | NP_956647.2 | 3 - 4 | 425 | 64 | |
GenBank mRNA | BC053173.1 | - | 3 - 4 | 396 | 64 | |
BC154331.1 | - | 3 - 4 | 388 | 64 | ||
EH433128.1 | - | 3 - 4 | 450 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |