Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: marveld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200529.1 | NP_956823.1 | 3 - 4 | 423 | 94 | |
GenBank mRNA | BC055662.1 | - | 3 - 4 | 423 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |