Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phactr4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200670.1 | NP_956964.1 | 8 - 9 | 1664 | 150 | |
GenBank mRNA | BC058052.1 | - | 8 - 9 | 1664 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |