Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phactr4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200670.1 | NP_956964.1 | 9 - 10 | 1948 | 96 | |
XM_005158332.4 | XP_005158389.1 | 8 - 9 | 2333 | 96 | ||
XM_005158333.4 | XP_005158390.1 | 8 - 9 | 2561 | 96 | ||
XM_005158334.4 | XP_005158391.1 | 10 - 11 | 2327 | 96 | ||
XM_005158335.4 | XP_005158392.1 | 7 - 8 | 2199 | 96 | ||
XM_009292432.3 | XP_009290707.1 | 10 - 11 | 2827 | 96 | ||
XM_009292434.3 | XP_009290709.1 | 9 - 10 | 2195 | 96 | ||
XM_021466650.1 | XP_021322325.1 | 10 - 11 | 2275 | 96 | ||
GenBank mRNA | BC058052.1 | - | 9 - 10 | 1948 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |