Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paqr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200710.2 | NP_957004.1 | 4 - 5 | 732 | 67 | |
GenBank mRNA | BC059466.1 | - | 4 - 5 | 639 | 67 | |
BC071435.1 | - | 4 - 5 | 732 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |