Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab5ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200970.1 | NP_957264.1 | 3 - 4 | 402 | 119 | |
GenBank mRNA | BC049057.1 | - | 3 - 4 | 402 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |