Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201005.1 | NP_957299.1 | 1 - 2 | 144 | 70 | |
GenBank mRNA | BC047847.1 | - | 1 - 2 | 144 | 70 | |
BC075751.1 | - | 1 - 2 | 108 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |