Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201072.2 | NP_957366.1 | 1 - 2 | 263 | 83 | |
GenBank mRNA | AY555037.1 | - | 1 - 2 | 251 | 83 | |
BC049315.1 | - | 1 - 2 | 251 | 83 | ||
BC062828.1 | - | 1 - 2 | 263 | 83 | ||
EH454794.1 | - | 1 - 2 | 280 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |