Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hpda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201167.1 | NP_957461.1 | 7 - 8 | 460 | 70 | |
XM_005171729.3 | XP_005171786.1 | 7 - 8 | 496 | 70 | ||
GenBank mRNA | AY398361.1 | - | 6 - 7 | 424 | 70 | |
BC046075.1 | - | 7 - 8 | 460 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |